Learn More
Medchemexpress LLC Boc-bipiperidine-ethynylbenzoic acid | 2308497-81-4 | 99.2% | 412.52 | 50 MG
SDP
Catalog No. 5000348283
Encompass_Preferred
Boc-bipiperidine-ethynylbenzoic acid is an alkyl/ether-based PROTAC linker used in the synthesis of ARD-61. This product also functions as a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-48283 Supplier Medchemexpress LLC Supplier No. HY13966150MG
Boc-bipiperidine-ethynylbenzoic acid is an alkyl/ether-based PROTAC linker used in the synthesis of ARD-61. This product also functions as a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups