Learn More
Medchemexpress LLC Boc-bipiperidine-ethynylbenzoic acid | 2308497-81-4 | 99.2% | C24H32N2O4 | 10MG
SDP
Catalog No. 5000261188
Encompass_Preferred
Boc-bipiperidine-ethynylbenzoic acid is a Boc-protected bipiperidine-containing ethynylbenzoic acid used as a PROTAC linker and click-chemistry building block for conjugation via alkyne chemistry. It is supplied as a high-purity solid for research use.
  • High purity (≈99.2%) suitable for research applications.
  • Boc-protected bipiperidine scaffold enabling linker synthesis.
  • Ethynyl handle for copper-catalyzed azide-alkyne cycloaddition.
  • Solid, white to off-white appearance for easy handling.
  • Available in small milligram pack sizes for synthesis and screening.
  • Store sealed, away from moisture and light; cold storage recommended.
Catalog No. 50-002-61188 Supplier Medchemexpress LLC Supplier No. HY13966110MG
Boc-bipiperidine-ethynylbenzoic acid is a Boc-protected bipiperidine-containing ethynylbenzoic acid used as a PROTAC linker and click-chemistry building block for conjugation via alkyne chemistry. It is supplied as a high-purity solid for research use.
  • High purity (≈99.2%) suitable for research applications.
  • Boc-protected bipiperidine scaffold enabling linker synthesis.
  • Ethynyl handle for copper-catalyzed azide-alkyne cycloaddition.
  • Solid, white to off-white appearance for easy handling.
  • Available in small milligram pack sizes for synthesis and screening.
  • Store sealed, away from moisture and light; cold storage recommended.