Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-bipiperidine-ethynylbenzoic acid | 2308497-81-4 | MFCD34578497 | 99.2% | 412.52 g/mol | C24H32N2O4 | 5 MG
SDP

Catalog No. 5000223294
Encompass_Preferred

Boc-bipiperidine-ethynylbenzoic acid is an alkyne-containing PROTAC linker and click-chemistry building block used for assembling bifunctional degraders and for copper-catalyzed azide-alkyne cycloaddition (CuAAC). Supplied as a purified small-molecule reagent, it is intended for synthetic chemistry and bioconjugation workflows.

  • Contains terminal alkyne for copper-catalyzed azide-alkyne cycloaddition.
  • High purity (≈99.2%) suitable for synthesis and analytical work.
  • Available in small research pack sizes for method development.
  • Stable as a solid when stored sealed, away from moisture and light.
  • Recommended storage in solvent: -80°C (6 months) or -20°C (1 month).
  • Useful as a PROTAC linker and bioconjugation building block.

Catalog No. 50-002-23294 Supplier Medchemexpress LLC Supplier No. HY1396615MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-bipiperidine-ethynylbenzoic acid is an alkyne-containing PROTAC linker and click-chemistry building block used for assembling bifunctional degraders and for copper-catalyzed azide-alkyne cycloaddition (CuAAC). Supplied as a purified small-molecule reagent, it is intended for synthetic chemistry and bioconjugation workflows.

  • Contains terminal alkyne for copper-catalyzed azide-alkyne cycloaddition.
  • High purity (≈99.2%) suitable for synthesis and analytical work.
  • Available in small research pack sizes for method development.
  • Stable as a solid when stored sealed, away from moisture and light.
  • Recommended storage in solvent: -80°C (6 months) or -20°C (1 month).
  • Useful as a PROTAC linker and bioconjugation building block.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.