Learn More
Medchemexpress LLC Boc-C1-PEG2-C4-Cl (PROTAC linker 1) | 1835705-53-7 | 99.8% | 294.81 | 250 MG
SDP
Catalog No. 5000347595
Encompass_Preferred
Boc-C1-PEG2-C4-Cl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTAC technology employs small molecules that recruit target proteins for ubiquitination and removal by the proteasome.
  • The synthesis of PROTAC compounds that mediate the degradation of c-ABL and BCR-ABL by recruiting either Cereblon or Von Hippel Lindau E3 ligases is reported.
  • Designated as 6-2-2.
Catalog No. 50-003-47595 Supplier Medchemexpress LLC Supplier No. 250MGHY108371
Boc-C1-PEG2-C4-Cl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTAC technology employs small molecules that recruit target proteins for ubiquitination and removal by the proteasome.
  • The synthesis of PROTAC compounds that mediate the degradation of c-ABL and BCR-ABL by recruiting either Cereblon or Von Hippel Lindau E3 ligases is reported.
  • Designated as 6-2-2.