Learn More
Medchemexpress LLC Boc-C1-PEG3-C4-OBn | 2381196-81-0 | 95.8% | 50 MG
SDP
Catalog No. 5000341101
Encompass_Preferred
Boc-C1-PEG3-C4-OBn is a linker compound that is characterized by its polyethylene glycol (PEG) composition. This product is utilized in the creation of various PROTACs, which are molecules designed with two distinct ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs harness the ubiquitin-proteasome system within cells to achieve selective degradation of target proteins.
  • This is a linker compound with a polyethylene glycol composition.
  • It facilitates the synthesis of various PROTAC molecules.
  • It contributes to the formation of PROTACs that can selectively degrade target proteins.
  • The product has a high purity of 95.77%.
Catalog No. 50-003-41101 Supplier Medchemexpress LLC Supplier No. HY13061950MG
Boc-C1-PEG3-C4-OBn is a linker compound that is characterized by its polyethylene glycol (PEG) composition. This product is utilized in the creation of various PROTACs, which are molecules designed with two distinct ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs harness the ubiquitin-proteasome system within cells to achieve selective degradation of target proteins.
  • This is a linker compound with a polyethylene glycol composition.
  • It facilitates the synthesis of various PROTAC molecules.
  • It contributes to the formation of PROTACs that can selectively degrade target proteins.
  • The product has a high purity of 95.77%.