Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-C1-PEG3-C4-OH | 2376724-97-7 | >95.0% | 50 MG
SDP

Catalog No. 5000341092
Encompass_Preferred

Boc-C1-PEG3-C4-OH is a PROTAC linker, appearing as a liquid. It has a molecular formula of C16H32O6 and a molecular weight of 320.42. Intended for laboratory chemicals and substance manufacturing, it is supplied in a quantity of 50 MG with a purity of >95.0%.

  • PROTAC linker
  • Molecular formula: C16H32O6
  • Molecular weight: 320.42
  • Appearance: Liquid
  • For laboratory chemical use
  • For substance manufacturing
  • Purity: >95.0%
  • Store pure form at -20°C for 3 years or 4°C for 2 years

Catalog No. 50-003-41092 Supplier Medchemexpress LLC Supplier No. HY13061850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-C1-PEG3-C4-OH is a PROTAC linker, appearing as a liquid. It has a molecular formula of C16H32O6 and a molecular weight of 320.42. Intended for laboratory chemicals and substance manufacturing, it is supplied in a quantity of 50 MG with a purity of >95.0%.

  • PROTAC linker
  • Molecular formula: C16H32O6
  • Molecular weight: 320.42
  • Appearance: Liquid
  • For laboratory chemical use
  • For substance manufacturing
  • Purity: >95.0%
  • Store pure form at -20°C for 3 years or 4°C for 2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.