Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-C14-COOH | 843666-27-3 | 97.0% | 342.51 g·mol⁻¹ | C20H38O4 | 10 G
SDP

Catalog No. 5000202852
Encompass_Preferred

Boc-C14-COOH is a Boc-protected C14 alkyl carboxylic acid used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as an alkyl-chain linker for PROTAC design.

  • Non-cleavable ADC linker for conjugation chemistry.
  • Alkyl-chain PROTAC linker for targeted protein degradation.
  • Molecular formula C20H38O4, molecular weight 342.51 g·mol⁻¹.
  • Purity typically 97.0% as supplied.
  • Available in laboratory pack sizes, including 10 G.

Catalog No. 50-002-02852 Supplier Medchemexpress LLC Supplier No. HYW03459910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-C14-COOH is a Boc-protected C14 alkyl carboxylic acid used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as an alkyl-chain linker for PROTAC design.

  • Non-cleavable ADC linker for conjugation chemistry.
  • Alkyl-chain PROTAC linker for targeted protein degradation.
  • Molecular formula C20H38O4, molecular weight 342.51 g·mol⁻¹.
  • Purity typically 97.0% as supplied.
  • Available in laboratory pack sizes, including 10 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.