Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-C14-COOH | 843666-27-3 | >98.0% | 342.51 | C20H38O4 | 100 G
SDP

Catalog No. 5000203228
Encompass_Preferred

Boc-C14-COOH is a Boc-protected C14 carboxylic acid used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as an alkyl-chain linker for proteolysis targeting chimera (PROTAC) construction. It is supplied as a solid, white to off-white powder suitable for conjugation chemistries.

  • Non-cleavable ADC linker for antibody conjugation
  • Alkyl chain-based PROTAC linker
  • Purity >98.0%
  • Molecular weight 342.51 g/mol
  • Chemical formula C20H38O4
  • Physical form: white to off-white solid
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years)

Catalog No. 50-002-03228 Supplier Medchemexpress LLC Supplier No. HYW034599100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-C14-COOH is a Boc-protected C14 carboxylic acid used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as an alkyl-chain linker for proteolysis targeting chimera (PROTAC) construction. It is supplied as a solid, white to off-white powder suitable for conjugation chemistries.

  • Non-cleavable ADC linker for antibody conjugation
  • Alkyl chain-based PROTAC linker
  • Purity >98.0%
  • Molecular weight 342.51 g/mol
  • Chemical formula C20H38O4
  • Physical form: white to off-white solid
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.