Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-C14-COOH | 843666-27-3 | ≥98.0% | 342.51 g/mol | C20H38O4 | 5 G
SDP

Catalog No. 5000203143
Encompass_Preferred

Boc-C14-COOH is a Boc-protected C14 carboxylic acid used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as an alkyl chain linker in PROTAC construction. It is supplied as a powder with characterized solubility and storage recommendations.

  • Used as a non-cleavable ADC linker and PROTAC linker.
  • Powder form with specified storage conditions for powder and solutions.
  • Soluble in DMSO (200 mg/mL) for in vitro use; formulations for in vivo use are documented.
  • Molecular weight 342.51 g/mol and formula C20H38O4 for formulation calculations.
  • Available in small laboratory quantities and larger pack sizes per vendor listing.

Catalog No. 50-002-03143 Supplier Medchemexpress LLC Supplier No. HYW0345995G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-C14-COOH is a Boc-protected C14 carboxylic acid used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as an alkyl chain linker in PROTAC construction. It is supplied as a powder with characterized solubility and storage recommendations.

  • Used as a non-cleavable ADC linker and PROTAC linker.
  • Powder form with specified storage conditions for powder and solutions.
  • Soluble in DMSO (200 mg/mL) for in vitro use; formulations for in vivo use are documented.
  • Molecular weight 342.51 g/mol and formula C20H38O4 for formulation calculations.
  • Available in small laboratory quantities and larger pack sizes per vendor listing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.