Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-C14-COOH | 843666-27-3 | ≥98.0% | 342.51 g/mol | C20H38O4 | 50 G
SDP

Catalog No. 5000203224
Encompass_Preferred

Boc-C14-COOH is a Boc-protected C14 alkyl carboxylic acid linker used as a non-cleavable spacer in the synthesis of antibody-drug conjugates (ADCs) and in PROTAC linker construction. It provides a hydrophobic tetradecyl spacer terminating in a protected carboxyl suitable for conjugation chemistry.

  • Non-cleavable C14 alkyl linker for ADC and PROTAC synthesis.
  • Boc-protected carboxylic acid functional group for conjugation.
  • Molecular weight 342.51 g/mol and chemical formula C20H38O4.
  • CAS number 843666-27-3.
  • Available as a 50 g powder.
  • Storage: powder -20°C for 3 years, 4°C for 2 years.

Catalog No. 50-002-03224 Supplier Medchemexpress LLC Supplier No. HYW03459950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-C14-COOH is a Boc-protected C14 alkyl carboxylic acid linker used as a non-cleavable spacer in the synthesis of antibody-drug conjugates (ADCs) and in PROTAC linker construction. It provides a hydrophobic tetradecyl spacer terminating in a protected carboxyl suitable for conjugation chemistry.

  • Non-cleavable C14 alkyl linker for ADC and PROTAC synthesis.
  • Boc-protected carboxylic acid functional group for conjugation.
  • Molecular weight 342.51 g/mol and chemical formula C20H38O4.
  • CAS number 843666-27-3.
  • Available as a 50 g powder.
  • Storage: powder -20°C for 3 years, 4°C for 2 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.