Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-C14-COOH | 843666-27-3 | MFCD23136039 | ≥98.0% | 342.51 g/mol | C20H38O4 | 25 G
SDP

Catalog No. 5000203112
Encompass_Preferred

Boc-protected C14 alkyl-chain carboxylic acid used as a non-cleavable linker in antibody-drug conjugate synthesis and as an alkyl-chain PROTAC linker. Supplied as a solid for research use.

  • Non-cleavable linker for ADC synthesis.
  • Suitable as an alkyl-chain PROTAC linker.
  • Purity ≥98.0% and molecular weight 342.51 g/mol.
  • Molecular formula C20H38O4.
  • Available as a 25 G solid for research applications.

Catalog No. 50-002-03112 Supplier Medchemexpress LLC Supplier No. HYW03459925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-protected C14 alkyl-chain carboxylic acid used as a non-cleavable linker in antibody-drug conjugate synthesis and as an alkyl-chain PROTAC linker. Supplied as a solid for research use.

  • Non-cleavable linker for ADC synthesis.
  • Suitable as an alkyl-chain PROTAC linker.
  • Purity ≥98.0% and molecular weight 342.51 g/mol.
  • Molecular formula C20H38O4.
  • Available as a 25 G solid for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.