Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-C16-COOH | 843666-40-0 | 100.0% | 370.57 g/mol | C22H42O4 | 1 G
SDP

Catalog No. 5000202928
Encompass_Preferred

Boc-C16-COOH is an alkyl-chain linker used as a non-cleavable component in antibody-drug conjugate (ADC) synthesis and as a linker building block for proteolysis-targeting chimeras (PROTACs). It is a small-molecule research reagent with formula C22H42O4, molecular weight 370.57 g/mol, and is supplied with high reported purity for biochemical research applications.

  • Alkyl-chain non-cleavable ADC linker.
  • Suitable as an alkyl spacer for PROTAC design.
  • Molecular formula C22H42O4.
  • Molecular weight 370.57 g/mol.
  • Reported CAS number 843666-40-0.
  • Available in small research pack sizes, including 1 G.

Catalog No. 50-002-02928 Supplier Medchemexpress LLC Supplier No. HYW0455981G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-C16-COOH is an alkyl-chain linker used as a non-cleavable component in antibody-drug conjugate (ADC) synthesis and as a linker building block for proteolysis-targeting chimeras (PROTACs). It is a small-molecule research reagent with formula C22H42O4, molecular weight 370.57 g/mol, and is supplied with high reported purity for biochemical research applications.

  • Alkyl-chain non-cleavable ADC linker.
  • Suitable as an alkyl spacer for PROTAC design.
  • Molecular formula C22H42O4.
  • Molecular weight 370.57 g/mol.
  • Reported CAS number 843666-40-0.
  • Available in small research pack sizes, including 1 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.