Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-C16-cooh | 843666-40-0 | 100.0% | 370.57 g/mol | C22H42O4 | 500 MG
SDP

Catalog No. 5000203117
Encompass_Preferred

Boc-C16-COOH is a Boc-protected long-chain carboxylic acid used as a non-cleavable antibody-drug conjugate (ADC) linker and as an alkyl-chain PROTAC linker for chemical synthesis. It is supplied as a white to off-white solid with high purity and is soluble in DMSO for stock solutions.

  • Non-cleavable ADC and PROTAC linker
  • High purity (99.98%)
  • Molecular weight 370.57 g/mol
  • Cas number 843666-40-0
  • Soluble in DMSO: 100 mg/mL; ultrasonic and gentle heating (to 60°C) may improve solubility
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Appearance: white to off-white solid
  • Available unit: 500 MG

Catalog No. 50-002-03117 Supplier Medchemexpress LLC Supplier No. HYW045598500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-C16-COOH is a Boc-protected long-chain carboxylic acid used as a non-cleavable antibody-drug conjugate (ADC) linker and as an alkyl-chain PROTAC linker for chemical synthesis. It is supplied as a white to off-white solid with high purity and is soluble in DMSO for stock solutions.

  • Non-cleavable ADC and PROTAC linker
  • High purity (99.98%)
  • Molecular weight 370.57 g/mol
  • Cas number 843666-40-0
  • Soluble in DMSO: 100 mg/mL; ultrasonic and gentle heating (to 60°C) may improve solubility
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Appearance: white to off-white solid
  • Available unit: 500 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.