Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-C2-Urea-bis(Boc)-C4-Urea-4-phenylacetic acid | 1610413-97-2 | 96.9% | 678.81 | 50 MG
SDP

Catalog No. 5000347166
Encompass_Preferred

Boc-C2-Urea-bis(Boc)-C4-Urea-4-phenylacetic acid is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs).

  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Cleavable ADC linker
  • Purity of 96.87%
  • Molecular weight of 678.81
  • Appears as a white to off-white solid
  • Soluble in DMSO (100 mg/mL)
  • Stable as powder for 3 years at -20°C
  • Stable in solvent for 6 months at -80°C

Catalog No. 50-003-47166 Supplier Medchemexpress LLC Supplier No. HY1083792MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-C2-Urea-bis(Boc)-C4-Urea-4-phenylacetic acid is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs).

  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Cleavable ADC linker
  • Purity of 96.87%
  • Molecular weight of 678.81
  • Appears as a white to off-white solid
  • Soluble in DMSO (100 mg/mL)
  • Stable as powder for 3 years at -20°C
  • Stable in solvent for 6 months at -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.