Learn More
Medchemexpress LLC Boc-C5-O-C5-O-C6-Cl | 1835705-52-6 | 393.00 | 1 MG
SDP
Catalog No. 502080741
Encompass_Preferred
Boc-C5-O-C5-O-C6-Cl is a PROTAC linker used to connect a tyrosine kinase inhibitor (TKI) to an E3 recruiting ligand.
  • Utilized for PROTAC design.
  • Connects tyrosine kinase inhibitors to E3 recruiting ligands.
Catalog No. 50-208-0741 Supplier Medchemexpress LLC Supplier No. HY108372100MG
Boc-C5-O-C5-O-C6-Cl is a PROTAC linker used to connect a tyrosine kinase inhibitor (TKI) to an E3 recruiting ligand.
  • Utilized for PROTAC design.
  • Connects tyrosine kinase inhibitors to E3 recruiting ligands.