Learn More
Bachem Boc-Cys(NPys)-OH
SDP
Catalog No. 50258381
Encompass_Preferred
Boc-Cys(NPys)-OH, 76880-29-0, C13H17N3O6S2, Mr 375.43, 5g. BCNP. Npys allows site-directed disulfide bond formation, as it is cleaved by thiols. Galande and Spatola obtained disulfide heterodimers of peptides via coupling Boc-Cys(NPys)-OH together with Fmoc-Cys(Mmt)-OH to MBHA-resin followed by Mmt removal with 1% TFA in dichloromethane. Mourier et al. used BCNP for the identification of T-cell epitopes in Cys and/or Cyt-containing proteins.
Catalog No. 50-258-381 Supplier Bachem Supplier No. 4014705.0005
Add to Cart
Add to Cart
Boc-Cys(NPys)-OH, 76880-29-0, C13H17N3O6S2, Mr 375.43, 5g. BCNP. Npys allows site-directed disulfide bond formation, as it is cleaved by thiols. Galande and Spatola obtained disulfide heterodimers of peptides via coupling Boc-Cys(NPys)-OH together with Fmoc-Cys(Mmt)-OH to MBHA-resin followed by Mmt removal with 1% TFA in dichloromethane. Mourier et al. used BCNP for the identification of T-cell epitopes in Cys and/or Cyt-containing proteins.