Learn More
AdipoGen Boc-Cys-OH2
SDP
Catalog No. 502561637
Encompass_Preferred
Chemical. CAS 10389-65-8. Formula C16H28N2O8S2. MW 440.53. Synthetic. Amino acid protected by tert-butoxycarbonyl Boc group. Used as a reactant in peptide synthesis. Used to obtain asymmetric cystine peptides by enzymatic catalysis with immobilized papain. Used as educt for obtaining S-isoacyl dipeptide building blocks.
Catalog No. 50-256-1637 Supplier AdipoGen Supplier No. CDXB0224G005
Chemical. CAS 10389-65-8. Formula C16H28N2O8S2. MW 440.53. Synthetic. Amino acid protected by tert-butoxycarbonyl Boc group. Used as a reactant in peptide synthesis. Used to obtain asymmetric cystine peptides by enzymatic catalysis with immobilized papain. Used as educt for obtaining S-isoacyl dipeptide building blocks.