Learn More
Medchemexpress LLC Boc-D-(4-fmoc)-aminophenylalanine | 173054-11-0 | 502.56 | 1 G
SDP
Catalog No. 5000460423
Encompass_Preferred
Boc-D-(4-fmoc)-aminophenylalanine is an amino acid derivative that contains an Fmoc protecting group. It can be utilized in the synthesis of peptides exhibiting gonadotropin-releasing hormone antagonist activity.
  • Amino acid derivative with an Fmoc protecting group.
  • Can be used to synthesize peptides with gonadotropin-releasing hormone antagonist activity.
  • For research use only.
Catalog No. 50-004-60423 Supplier Medchemexpress LLC Supplier No. HYW0069371G
Boc-D-(4-fmoc)-aminophenylalanine is an amino acid derivative that contains an Fmoc protecting group. It can be utilized in the synthesis of peptides exhibiting gonadotropin-releasing hormone antagonist activity.
  • Amino acid derivative with an Fmoc protecting group.
  • Can be used to synthesize peptides with gonadotropin-releasing hormone antagonist activity.
  • For research use only.