Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-D-alaninol | 106391-86-0 | 175.23 | 100 G
SDP

Catalog No. 5000448731
Encompass_Preferred

Boc-D-alaninol is a chemical compound commonly used as a laboratory chemical and in the manufacture of various substances. It appears as a white to off-white solid.

  • Appears as a white to off-white solid
  • High purity of 99.9%
  • Low water content of 0.07%
  • Minimal residue on ignition (<0.01%)
  • Primarily for research use only
  • Identified for use in laboratory and substance manufacturing applications

Catalog No. 50-004-48731 Supplier Medchemexpress LLC Supplier No. HYW008185100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-D-alaninol is a chemical compound commonly used as a laboratory chemical and in the manufacture of various substances. It appears as a white to off-white solid.

  • Appears as a white to off-white solid
  • High purity of 99.9%
  • Low water content of 0.07%
  • Minimal residue on ignition (<0.01%)
  • Primarily for research use only
  • Identified for use in laboratory and substance manufacturing applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.