Learn More
Medchemexpress LLC Boc-D-alaninol | 106391-86-0 | MFCD00235912 | 175.23 | 25 G

Supplier: Medchemexpress LLC 25GHYW008185
Boc-D-alaninol is a chemical compound (C8H17NO3, MW 175.23) that appears as a white to off-white solid with a purity of 99.9%. It is classified as a laboratory chemical and is stable under recommended storage conditions.
- Used in the manufacture of substances
- White to off-white solid appearance
- High purity (99.9%)
- Stable under recommended storage conditions
- For research use only
- Melting point: 58-61°C
- Boiling point: 276.4°C at 760 mmHg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.