Learn More
Medchemexpress LLC Boc-D-Arg(Pbf)-OH | 186698-61-3 | 99.3% | 526.65 g/mol | C24H38N4O7S | 10 G
SDP
Catalog No. 5000210668
Encompass_Preferred
Boc-D-Arg(Pbf)-OH is a Boc-protected D-arginine derivative used as a building block in peptide synthesis. It contains a Pbf-protected guanidino side chain and a Boc-protected N-terminus, and is supplied as a high-purity solid for research use.
  • Boc-protected D-arginine for peptide assembly.
  • Pbf side-chain protection for guanidino group.
  • High purity suitable for analytical and synthetic use.
  • Solid form convenient for weighing and storage.
  • Intended for research use only.
Catalog No. 50-002-10668 Supplier Medchemexpress LLC Supplier No. HYW06230410G
Boc-D-Arg(Pbf)-OH is a Boc-protected D-arginine derivative used as a building block in peptide synthesis. It contains a Pbf-protected guanidino side chain and a Boc-protected N-terminus, and is supplied as a high-purity solid for research use.
  • Boc-protected D-arginine for peptide assembly.
  • Pbf side-chain protection for guanidino group.
  • High purity suitable for analytical and synthetic use.
  • Solid form convenient for weighing and storage.
  • Intended for research use only.