Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boc-D-Bpa-OH, Advanced ChemTech

Catalog No. 01341199
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
01-341-199 5 g
01-341-200 1 g
2 options

Catalog No. 01-341-199

Supplier: Advanced Chem Tech BF34245G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117666-94-1
Molecular Formula C21H23NO5
Molecular Weight (g/mol) 369.42
MDL Number MFCD00237572
InChI Key HIQJNYPOWPXYIC-QGZVFWFLSA-N
IUPAC Name (2R)-3-(4-benzoylphenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)C(O)=O

Specifications

Color White to Off-White
Quantity 5 g
Grade Biochemical
Chemical Name or Material Boc-D-Bpa-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.