Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-D-cyclopropylglycine | 609768-49-2 | C10H17NO4 | 1 G
SDP

Catalog No. 5000434773
Encompass_Preferred

Boc-D-cyclopropylglycine is a chemical compound designated for research and development applications only, not for medicinal, household, or other uses. This compound exhibits specific physical properties, including a boiling point of 364.7°C, a flash point of 174.4°C, and a density of 1.206 g/cm3. Safe handling practices involve working in a well-ventilated environment, utilizing appropriate personal protective equipment, and storing the substance in a cool, dry, and well-ventilated area. Responsible disposal is crucial to prevent environmental contamination.

  • Intended for R&D use only
  • Boiling point: 364.7°C
  • Flash point: 174.4°C
  • Density: 1.206 g/cm3
  • Handle in well-ventilated area
  • Wear safety goggles and impervious gloves
  • Store in dry, cool, well-ventilated place
  • Dispose via chemical destruction or controlled incineration

Catalog No. 50-004-34773 Supplier Medchemexpress LLC Supplier No. HY602651G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-D-cyclopropylglycine is a chemical compound designated for research and development applications only, not for medicinal, household, or other uses. This compound exhibits specific physical properties, including a boiling point of 364.7°C, a flash point of 174.4°C, and a density of 1.206 g/cm3. Safe handling practices involve working in a well-ventilated environment, utilizing appropriate personal protective equipment, and storing the substance in a cool, dry, and well-ventilated area. Responsible disposal is crucial to prevent environmental contamination.

  • Intended for R&D use only
  • Boiling point: 364.7°C
  • Flash point: 174.4°C
  • Density: 1.206 g/cm3
  • Handle in well-ventilated area
  • Wear safety goggles and impervious gloves
  • Store in dry, cool, well-ventilated place
  • Dispose via chemical destruction or controlled incineration

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.