Learn More
Apexbio Technology LLC Boc-D-FMK, 5mg. Cas: 187389-53-3,634911-80-1 MFCD: MFCD03453073. Pan-caspase inhibitor
SDP
Supplier:  Apexbio Technology LLC A19045
Encompass_Preferred
Boc-D-FMK is a cell-permeable broad-spectrum caspase inhibitor that fully inhibits the pro-apoptotic effect of tumor necrosis factor- (TNF) with the half maximal inhibition concentration IC50 value of 39 M [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4721