Learn More
Medchemexpress LLC HY-13229 5mg Medchemexpress, BOC-D-FMK CAS:634911-80-1 Purity:>98%
SDP
Catalog No. 502081308
Encompass_Preferred
Medchemexpress, HY-13229 5mg BOC-D-FMK CAS:634911-80-1 Boc-D-FMK is a cell-permeable, irreversible and broad spectrum caspase inhibitor. Boc-D-FMK inhibits apoptosis stimulated by TNF-α with an IC50 of 39 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1308 Supplier Medchemexpress LLC Supplier No. HY132295MG
Medchemexpress, HY-13229 5mg BOC-D-FMK CAS:634911-80-1 Boc-D-FMK is a cell-permeable, irreversible and broad spectrum caspase inhibitor. Boc-D-FMK inhibits apoptosis stimulated by TNF-α with an IC50 of 39 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.