Learn More
Abcam Boc-D-FMK, caspase inhibitor, 5MG
Catalog No. 502706369
Encompass_Preferred
MW 263.26 Da. Irreversible general caspase inhibitor. Blocks hepatocyte apoptosis in vivo. Active in vitro and in vivo. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6369 Supplier Abcam Supplier No. AB1420365MG
MW 263.26 Da. Irreversible general caspase inhibitor. Blocks hepatocyte apoptosis in vivo. Active in vitro and in vivo. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement