Learn More
Medchemexpress LLC Boc-D-homoserine | 745011-75-0 | MFCD12545888 | 98.0% | 219.24 | C9H17NO5 | 25 G
SDP
Catalog No. 5000303074
Encompass_Preferred
Boc-D-homoserine is the N-Boc (tert-butoxycarbonyl) protected D-enantiomer of homoserine supplied as a white to off-white solid for research use. It functions as an amino acid derivative and versatile synthetic building block in peptide and organic synthesis. Typical specifications include CAS 745011-75-0, molecular formula C9H17NO5, molecular weight 219.24 Da, and listed purity of 98.0%.
  • Versatile building block for peptide and small-molecule synthesis.
  • N-Boc protection enables selective amine chemistry.
  • High purity (98.0%) suitable for research applications.
  • Solid powder form for convenient handling and storage.
  • Available in multiple pack sizes for scale flexibility.
Catalog No. 50-003-03074 Supplier Medchemexpress LLC Supplier No. 25GHYW017788
Boc-D-homoserine is the N-Boc (tert-butoxycarbonyl) protected D-enantiomer of homoserine supplied as a white to off-white solid for research use. It functions as an amino acid derivative and versatile synthetic building block in peptide and organic synthesis. Typical specifications include CAS 745011-75-0, molecular formula C9H17NO5, molecular weight 219.24 Da, and listed purity of 98.0%.
  • Versatile building block for peptide and small-molecule synthesis.
  • N-Boc protection enables selective amine chemistry.
  • High purity (98.0%) suitable for research applications.
  • Solid powder form for convenient handling and storage.
  • Available in multiple pack sizes for scale flexibility.