Learn More
Medchemexpress LLC Boc-D-HoPro-OH | 28697-17-8 | 98.0% | 229.28 | 25 G
SDP
Catalog No. 5000451401
Encompass_Preferred
Boc-D-HoPro-OH is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also applied as a catalytic agent and petrochemical additive, and is an important organic intermediate used in agrochemicals, pharmaceuticals, and dyestuff fields.
  • Can be used as a biological material
  • Can be used as an organic compound for life science related research
  • Applied as a catalytic agent
  • Applied as a petrochemical additive
  • Important organic intermediate in agrochemicals, pharmaceuticals, and dyestuff fields
Catalog No. 50-004-51401 Supplier Medchemexpress LLC Supplier No. 25GHYW013473
Boc-D-HoPro-OH is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also applied as a catalytic agent and petrochemical additive, and is an important organic intermediate used in agrochemicals, pharmaceuticals, and dyestuff fields.
  • Can be used as a biological material
  • Can be used as an organic compound for life science related research
  • Applied as a catalytic agent
  • Applied as a petrochemical additive
  • Important organic intermediate in agrochemicals, pharmaceuticals, and dyestuff fields