Learn More
Medchemexpress LLC Boc-D-Lys-OH | 106719-44-2 | 98.0% | 246.30 | 25 G
SDP
Catalog No. 5000452672
Encompass_Preferred
Boc-D-Lys-OH is a lysine derivative used for research purposes. It is a solid, white to off-white in appearance, with a molecular weight of 246.30 and a chemical formula of C11H22N2O4. This product is suitable for various laboratory applications.
  • Lysine derivative
  • Solid, white to off-white appearance
  • Available in various sizes
  • Used as an ergogenic supplement impacting anabolic hormone secretion and energy supply during exercise
  • Helps in preventing exercise-induced muscle damage
Catalog No. 50-004-52672 Supplier Medchemexpress LLC Supplier No. 25GHYW011977
Boc-D-Lys-OH is a lysine derivative used for research purposes. It is a solid, white to off-white in appearance, with a molecular weight of 246.30 and a chemical formula of C11H22N2O4. This product is suitable for various laboratory applications.
  • Lysine derivative
  • Solid, white to off-white appearance
  • Available in various sizes
  • Used as an ergogenic supplement impacting anabolic hormone secretion and energy supply during exercise
  • Helps in preventing exercise-induced muscle damage