Learn More
Medchemexpress LLC Boc-D-norleucine (Boc-D-Nle-OH) | 55674-63-0 | 98.0% | 231.29 | 10 G
SDP
Catalog No. 5000404940
Encompass_Preferred
Boc-D-norleucine (Boc-D-Nle-OH) is a leucine derivative that is utilized in peptide synthesis. It is intended for research use only and is not sold to patients.
  • Leucine derivative
  • Can be used for peptide synthesis
  • Available in various quantities from 1g to 100g
Catalog No. 50-004-04940 Supplier Medchemexpress LLC Supplier No. HY41912B10G
Boc-D-norleucine (Boc-D-Nle-OH) is a leucine derivative that is utilized in peptide synthesis. It is intended for research use only and is not sold to patients.
  • Leucine derivative
  • Can be used for peptide synthesis
  • Available in various quantities from 1g to 100g