Learn More
Medchemexpress LLC Boc-D-Phe(4-Cl)-OH | 57292-44-1 | MFCD00076978 | 100.0% | 299.75 | C14H18ClNO4 | 100 G
SDP
Catalog No. 5000300581
Encompass_Preferred
Boc-D-Phe(4-Cl)-OH is a Boc-protected D-phenylalanine derivative with a 4-chloro substitution on the aromatic ring, provided as a purified powder for research use. It is used as a stereochemically defined building block in peptide synthesis and related organic chemistry applications, offering reliable performance where a protected D-amino acid is required.
  • Boc-protected D-phenylalanine derivative with 4-chloro substitution.
  • Suitable as a building block for peptide and organic synthesis.
  • Supplied as a stable powder for convenient handling and storage.
  • High purity (99.97%) for consistent experimental results.
  • Offered in multiple package sizes to support small-scale and larger preparations.
Catalog No. 50-003-00581 Supplier Medchemexpress LLC Supplier No. HYW016018100G
Boc-D-Phe(4-Cl)-OH is a Boc-protected D-phenylalanine derivative with a 4-chloro substitution on the aromatic ring, provided as a purified powder for research use. It is used as a stereochemically defined building block in peptide synthesis and related organic chemistry applications, offering reliable performance where a protected D-amino acid is required.
  • Boc-protected D-phenylalanine derivative with 4-chloro substitution.
  • Suitable as a building block for peptide and organic synthesis.
  • Supplied as a stable powder for convenient handling and storage.
  • High purity (99.97%) for consistent experimental results.
  • Offered in multiple package sizes to support small-scale and larger preparations.