Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-D-phenylalanyl-D-phenylalanine | 125511-90-2 | 99.6% | 412.48 g/mol | C23H28N2O5 | 5 G
SDP

Catalog No. 5000280591
Encompass_Preferred

Boc-D-phenylalanyl-D-phenylalanine is a Boc-protected D-dipeptide supplied as a research-grade biochemical reagent for peptide synthesis and life-science assays. It is provided as a powder for laboratory use in organic synthesis and biochemical studies.

  • Boc-protected D-dipeptide suitable for peptide synthesis and assays.
  • High purity (99.6% LCMS).
  • Molecular weight 412.48 g/mol.
  • Chemical formula C23H28N2O5.
  • Available in pre-weighed packages: 1 G, 5 G, 10 G, 25 G.

Catalog No. 50-002-80591 Supplier Medchemexpress LLC Supplier No. HYW2501405G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-D-phenylalanyl-D-phenylalanine is a Boc-protected D-dipeptide supplied as a research-grade biochemical reagent for peptide synthesis and life-science assays. It is provided as a powder for laboratory use in organic synthesis and biochemical studies.

  • Boc-protected D-dipeptide suitable for peptide synthesis and assays.
  • High purity (99.6% LCMS).
  • Molecular weight 412.48 g/mol.
  • Chemical formula C23H28N2O5.
  • Available in pre-weighed packages: 1 G, 5 G, 10 G, 25 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.