Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-D-Pyr-Oet | 144978-35-8 | 257.28 | 50 G
SDP

Catalog No. 5000445646
Encompass_Preferred

Boc-D-Pyr-Oet (N-Boc-D-pyroglutamic acid ethyl ester) is a biochemical assay reagent. This compound has a purity of 99.97% and is intended for research use only.

  • High purity: 99.97%
  • Molecular weight: 257.28
  • Chemical formula: C12H19NO5
  • CAS number: 144978-35-8
  • Appearance: Solid
  • Color: White to off-white
  • Stable at room temperature for 3 years
  • Stable in solvent at -80°C for 2 years, or -20°C for 1 year
  • Soluble in DMSO at 200 mg/mL

Catalog No. 50-004-45646 Supplier Medchemexpress LLC Supplier No. HYW00360550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-D-Pyr-Oet (N-Boc-D-pyroglutamic acid ethyl ester) is a biochemical assay reagent. This compound has a purity of 99.97% and is intended for research use only.

  • High purity: 99.97%
  • Molecular weight: 257.28
  • Chemical formula: C12H19NO5
  • CAS number: 144978-35-8
  • Appearance: Solid
  • Color: White to off-white
  • Stable at room temperature for 3 years
  • Stable in solvent at -80°C for 2 years, or -20°C for 1 year
  • Soluble in DMSO at 200 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.