Learn More
Medchemexpress LLC Boc-D-Ser(tBu)-OH | 248921-66-6 | MFCD09751020 | 97.0% | 261.32 g/mol | C12H23NO5 | 50 G
SDP
Catalog No. 5000301314
Encompass_Preferred
Boc-D-Ser(tBu)-OH is an N-Boc, O-tert-butyl protected D-serine derivative used as a building block in peptide synthesis and organic synthesis. It provides orthogonal protection for the amino and hydroxyl groups, enabling selective deprotection and coupling steps during multi-step syntheses.
  • N-Boc and O-tert-butyl protection for orthogonal deprotection strategies.
  • D-configuration for stereospecific synthesis applications.
  • High purity suitable for research use (97.0%).
  • Powder form with defined storage recommendations to preserve stability.
  • Commonly used as a protected amino acid intermediate in peptide and small-molecule synthesis.
  • Available in research-scale quantities including 50 g packs.
Catalog No. 50-003-01314 Supplier Medchemexpress LLC Supplier No. HYW01836650G
Boc-D-Ser(tBu)-OH is an N-Boc, O-tert-butyl protected D-serine derivative used as a building block in peptide synthesis and organic synthesis. It provides orthogonal protection for the amino and hydroxyl groups, enabling selective deprotection and coupling steps during multi-step syntheses.
  • N-Boc and O-tert-butyl protection for orthogonal deprotection strategies.
  • D-configuration for stereospecific synthesis applications.
  • High purity suitable for research use (97.0%).
  • Powder form with defined storage recommendations to preserve stability.
  • Commonly used as a protected amino acid intermediate in peptide and small-molecule synthesis.
  • Available in research-scale quantities including 50 g packs.