Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-Dap(Boc)-OH | 88971-40-8 | ≥95.0% | 304.34 g/mol | C13H24N2O6 | 500 MG
SDP

Catalog No. 5000210879
Encompass_Preferred

Boc-Dap(Boc)-OH is a Boc-protected 2,3-diaminopropionic acid derivative used as a building block in peptide synthesis and medicinal chemistry research. It is supplied as a solid research chemical for synthesis and analytical applications.

  • Boc-protected diamino acid for peptide synthesis.
  • CAS number 88971-40-8.
  • Molecular formula C13H24N2O6; molecular weight 304.34 g/mol.
  • Purity ≥95.0% as stated on the certificate of analysis.
  • Physical form: solid (white to off-white).
  • Available in laboratory pack sizes including 500 mg.
  • Storage: store at room temperature for long-term; in solution, store at -20°C to -80°C.

Catalog No. 50-002-10879 Supplier Medchemexpress LLC Supplier No. HYW047799500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Dap(Boc)-OH is a Boc-protected 2,3-diaminopropionic acid derivative used as a building block in peptide synthesis and medicinal chemistry research. It is supplied as a solid research chemical for synthesis and analytical applications.

  • Boc-protected diamino acid for peptide synthesis.
  • CAS number 88971-40-8.
  • Molecular formula C13H24N2O6; molecular weight 304.34 g/mol.
  • Purity ≥95.0% as stated on the certificate of analysis.
  • Physical form: solid (white to off-white).
  • Available in laboratory pack sizes including 500 mg.
  • Storage: store at room temperature for long-term; in solution, store at -20°C to -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.