Learn More
Medchemexpress LLC Boc-Dap-NE | 160800-65-7 | 99.9% | 420.54 | C23H36N2O5 | 25 MG
SDP
Catalog No. 5000204225
Encompass_Preferred
Boc-Dap-NE is a Boc-protected dipeptide intermediate used in the synthesis of Monomethyl auristatin E (MMAE). It is supplied as a high-purity solid for research and development in antibody-drug conjugate (ADC) payload and linker synthesis.
  • High purity: 99.9%.
  • Molecular formula C23H36N2O5, molecular weight 420.54 g/mol.
  • Offered as a 25 MG solid, with larger sizes available.
  • Suitable for use as an intermediate in MMAE and ADC payload synthesis.
Catalog No. 50-002-04225 Supplier Medchemexpress LLC Supplier No. HY7893125MG
Boc-Dap-NE is a Boc-protected dipeptide intermediate used in the synthesis of Monomethyl auristatin E (MMAE). It is supplied as a high-purity solid for research and development in antibody-drug conjugate (ADC) payload and linker synthesis.
  • High purity: 99.9%.
  • Molecular formula C23H36N2O5, molecular weight 420.54 g/mol.
  • Offered as a 25 MG solid, with larger sizes available.
  • Suitable for use as an intermediate in MMAE and ADC payload synthesis.