Learn More
Medchemexpress LLC Boc-Dap-NE | 160800-65-7 | 99.9% | C23H36N2O5 | 25 MG
SDP
Catalog No. 5000358135
Encompass_Preferred
Boc-Dap-NE is an intermediate used in the synthesis of Monomethyl auristatin E (MMAE), a potent inhibitor of tubulin polymerization. It is commonly employed to create Antibody-Drug Conjugates (ADCs) as an ADC Cytotoxin.
  • Intermediate in Monomethyl auristatin E synthesis
  • Inhibits tubulin polymerization
  • Used to synthesize antibody-drug conjugates (ADCs)
  • High purity (99.9%)
  • Available documentation includes Data Sheet, COA, SDS, Handling Instructions, HNMR, RP-HPLC, NP-HPLC, LCMS
  • Solid, white to off-white appearance
Catalog No. 50-003-58135 Supplier Medchemexpress LLC Supplier No. 25MGHY78931A
Boc-Dap-NE is an intermediate used in the synthesis of Monomethyl auristatin E (MMAE), a potent inhibitor of tubulin polymerization. It is commonly employed to create Antibody-Drug Conjugates (ADCs) as an ADC Cytotoxin.
  • Intermediate in Monomethyl auristatin E synthesis
  • Inhibits tubulin polymerization
  • Used to synthesize antibody-drug conjugates (ADCs)
  • High purity (99.9%)
  • Available documentation includes Data Sheet, COA, SDS, Handling Instructions, HNMR, RP-HPLC, NP-HPLC, LCMS
  • Solid, white to off-white appearance