Learn More
Bachem Boc-Glu(OBzl)-Ala-Arg-AMC · HCl
SDP
Catalog No. 501946930
Encompass_Preferred
Boc-Glu(OBzl)-Ala-Arg-AMC · HCl, 133448-25-6, C36H47N7O9 · HCl, Mr 758.27, 250mg. Boc-EAR-AMC, fluorogenic substrate for coagulation factor XIa and trypsin.
Catalog No. 50-194-6930 Supplier Bachem Supplier No. 4017302.0250
Add to Cart
Add to Cart
Boc-Glu(OBzl)-Ala-Arg-AMC · HCl, 133448-25-6, C36H47N7O9 · HCl, Mr 758.27, 250mg. Boc-EAR-AMC, fluorogenic substrate for coagulation factor XIa and trypsin.