Learn More
Bachem Boc-Glu-Lys-Lys-AMC
SDP
Catalog No. 501946932
Encompass_Preferred
Boc-Glu-Lys-Lys-AMC Acetate, 73554-85-5, C32H48N6O9, Mr 660.77, 100mg. Boc-EKK-AMC, sensitive fluorogenic substrate for urokinase-activated plasmin.
Catalog No. 50-194-6932 Supplier Bachem Supplier No. 4007739.0100
Boc-Glu-Lys-Lys-AMC Acetate, 73554-85-5, C32H48N6O9, Mr 660.77, 100mg. Boc-EKK-AMC, sensitive fluorogenic substrate for urokinase-activated plasmin.