Learn More
Medchemexpress LLC Boc-Gln-Ala-Arg-AMC (acetate) | 00-00-0 | 99.7% | 690.74 g/mol | C31H46N8O10 | 5 MG
SDP
Catalog No. 5000222950
Encompass_Preferred
Boc-Gln-Ala-Arg-AMC acetate is an AMC-labeled peptide substrate for fluorescence-based protease assays. Enzymatic cleavage liberates the AMC fluorophore, producing a measurable fluorescent signal used in activity assays and screening applications.
  • Fluorogenic peptide substrate for protease activity assays.
  • Releases fluorescent AMC upon enzymatic hydrolysis.
  • Validated for trypsin and TMPRSS2 activity measurement.
  • High purity (99.7%) by HPLC.
  • Available in small pack sizes suitable for assay development.
Catalog No. 50-002-22950 Supplier Medchemexpress LLC Supplier No. HY134432B5MG
Boc-Gln-Ala-Arg-AMC acetate is an AMC-labeled peptide substrate for fluorescence-based protease assays. Enzymatic cleavage liberates the AMC fluorophore, producing a measurable fluorescent signal used in activity assays and screening applications.
  • Fluorogenic peptide substrate for protease activity assays.
  • Releases fluorescent AMC upon enzymatic hydrolysis.
  • Validated for trypsin and TMPRSS2 activity measurement.
  • High purity (99.7%) by HPLC.
  • Available in small pack sizes suitable for assay development.