Learn More
Bachem Boc-Gln-Ala-Arg-AMC · HCl
SDP
Catalog No. 50260323
Encompass_Preferred
Boc-Gln-Ala-Arg-AMC · HCl, 201849-55-0, C29H42N8O8 · HCl, Mr 667.16, 5mg. Boc-QAR-AMC, highly reactive substrate for trypsin. Fluorogenic substrate for prostasin and matriptase.
Catalog No. 50-260-323 Supplier Bachem Supplier No. 4017019.0005
Boc-Gln-Ala-Arg-AMC · HCl, 201849-55-0, C29H42N8O8 · HCl, Mr 667.16, 5mg. Boc-QAR-AMC, highly reactive substrate for trypsin. Fluorogenic substrate for prostasin and matriptase.