Learn More
Medchemexpress LLC Boc-Gln-Arg-Arg-AMC acetate | 2223665-74-3 | 99.25% | 775.85 | 10 MG
SDP
Catalog No. 5000397663
Encompass_Preferred
Boc-Gln-Arg-Arg-AMC acetate is a fluorogenic substrate utilized for determining protease activity. Upon hydrolysis, it releases the fluorescent product 7-amino-4-methylcoumarin (AMC), which emits a fluorescent signal under UV light. This product is for research use only.
  • Fluorogenic substrate
  • Used for determining protease activity
  • Releases fluorescent product 7-amino-4-methylcoumarin (AMC) upon hydrolysis
  • Emits fluorescent signal under UV light
Catalog No. 50-003-97663 Supplier Medchemexpress LLC Supplier No. HYP4323A10MG
Boc-Gln-Arg-Arg-AMC acetate is a fluorogenic substrate utilized for determining protease activity. Upon hydrolysis, it releases the fluorescent product 7-amino-4-methylcoumarin (AMC), which emits a fluorescent signal under UV light. This product is for research use only.
  • Fluorogenic substrate
  • Used for determining protease activity
  • Releases fluorescent product 7-amino-4-methylcoumarin (AMC) upon hydrolysis
  • Emits fluorescent signal under UV light