Learn More
Carbosynth LLC. Boc-Gln-Gly-Arg-MCA (5 mg vial), 1ea, MW: 616.67, Molecular formula: C28H40N8O8, CAS# 113865-85-3
SDP
Supplier:  Carbosynth LLC. MQR3136V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: t-Butyloxycarbonyl-L-Glutaminyl-Glycyl-L-Arginine 4-Methyl-Coumaryl-7-Amide, Boc-Gln-Gly-Arg-AMC, Boc-QGR-AMC, Boc-QGR-MCA, Purity: , Grade A, Application: Substrate for Factor Xlla, Storage Temperature: -20C, References: S. Kawabata, T. Miura, T. Morita, H. Kato, K. Fujikawa, S. Iwanaga, K. Takada, T. Kimura, and S. Sakakibara, Eur. J. Biochem., 172, 17 (1988).
Catalog No. 50-168-6018