Learn More
Cayman Chemical Boc-Glu-OBzl 1g
SDP
Catalog No. 502990619
Encompass_Preferred
An amino acid building block; has been used in the synthesis of peptide-based inhibitors of human caspases and HRV 3C protease that have enzyme inhibitory activity in vitro
Catalog No. 50-299-0619 Supplier Cayman Chemical Supplier No. 304641 G
An amino acid building block; has been used in the synthesis of peptide-based inhibitors of human caspases and HRV 3C protease that have enzyme inhibitory activity in vitro