Learn More
Cayman Chemical Boc-Glu-OBzl, Purity: ≥98%, CAS: 30924-93-7, 5 g
SDP
Catalog No. 502991748
Encompass_Preferred
An amino acid building block; has been used in the synthesis of peptide-based inhibitors of human caspases and HRV 3C protease that have enzyme inhibitory activity in vitro
Catalog No. 50-299-1748 Supplier Cayman Chemical Supplier No. 304645 G
An amino acid building block; has been used in the synthesis of peptide-based inhibitors of human caspases and HRV 3C protease that have enzyme inhibitory activity in vitro