Learn More
Carbosynth LLC. Boc-Gly-Arg-Arg-MCA (5 mg vial), 1ea, MW: 644.72, Molecular formula: C29H44N10O7, CAS# 113866-14-1
SDP
Supplier:  Carbosynth LLC. MGR3142V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: t-Butyloxycarbonyl-Glycyl-L-Arginyl-L-Arginine 4-Methyl-Coumaryl-7-Amide, Boc-Gly-Arg-Arg-AMC, Purity: , Grade A, Application: Substrate for Carboxyl Side of Paired Basic Residue Cleaving Enzyme, Storage Temperature: -20C, References: K. Mizuno, T. Nakamura, K. Takada, S. Sakakibara, and H. Matsuo, Biochem. Biophys. Res Commun., 144, 807 (1987).
Catalog No. 50-168-5980