Learn More
Medchemexpress LLC Boc-Gly-Gly-Phe-Gly-OH | 187794-49-6 | 99.4% | 436.46 | 10 G
SDP
Catalog No. 5000424392
Encompass_Preferred
Boc-Gly-Gly-Phe-Gly-OH, a self-assembly of N- and C-protected tetrapeptide, is a protease cleavable linker used for antibody-drug conjugates (ADCs).
  • Protease cleavable linker
  • Self-assembly of N- and C-protected tetrapeptide
  • Used for antibody-drug conjugate applications
  • Appears as a white to off-white solid
Catalog No. 50-004-24392 Supplier Medchemexpress LLC Supplier No. HYP144910G
Boc-Gly-Gly-Phe-Gly-OH, a self-assembly of N- and C-protected tetrapeptide, is a protease cleavable linker used for antibody-drug conjugates (ADCs).
  • Protease cleavable linker
  • Self-assembly of N- and C-protected tetrapeptide
  • Used for antibody-drug conjugate applications
  • Appears as a white to off-white solid