Learn More
Medchemexpress LLC Boc-Gly-Gly-Phe-Gly-OH | 187794-49-6 | 99.6% | 436.46 | 250 MG
SDP
Catalog No. 5000426747
Encompass_Preferred
Boc-Gly-Gly-Phe-Gly-OH is a self-assembly of N- and C-protected tetrapeptide. It functions as a protease cleavable linker specifically utilized in antibody-drug conjugates (ADC).
  • Self-assembly of N- and C-protected tetrapeptide
  • Protease cleavable linker
  • Used for antibody-drug conjugates
Catalog No. 50-004-26747 Supplier Medchemexpress LLC Supplier No. 250MGHYP1449
Boc-Gly-Gly-Phe-Gly-OH is a self-assembly of N- and C-protected tetrapeptide. It functions as a protease cleavable linker specifically utilized in antibody-drug conjugates (ADC).
  • Self-assembly of N- and C-protected tetrapeptide
  • Protease cleavable linker
  • Used for antibody-drug conjugates