Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-Gly-Lys-Arg-AMC | 109358-48-7 | 616.71 | 10 MG
SDP

Catalog No. 5000396728
Encompass_Preferred

Boc-Gly-Lys-Arg-AMC is a polypeptide used in peptide screening, a research tool that identifies active peptides primarily through immunoassay. This product is suitable for protein interaction studies, functional analysis, and epitope screening, particularly in agent research and development. It is intended for research use only.

  • Polypeptide for peptide screening
  • Used in protein interaction studies
  • Suitable for functional analysis
  • Ideal for epitope screening
  • Research use only
  • Chemical formula: C29H44N8O7
  • Sequence: {Boc-Gly}-Lys-{Arg-AMC}
  • Molecular weight: 616.71

Catalog No. 50-003-96728 Supplier Medchemexpress LLC Supplier No. HYP4314A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Gly-Lys-Arg-AMC is a polypeptide used in peptide screening, a research tool that identifies active peptides primarily through immunoassay. This product is suitable for protein interaction studies, functional analysis, and epitope screening, particularly in agent research and development. It is intended for research use only.

  • Polypeptide for peptide screening
  • Used in protein interaction studies
  • Suitable for functional analysis
  • Ideal for epitope screening
  • Research use only
  • Chemical formula: C29H44N8O7
  • Sequence: {Boc-Gly}-Lys-{Arg-AMC}
  • Molecular weight: 616.71

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.