Learn More
Enzo Life Sciences Boc-Gly-Val-Val-CHO (25mg)
SDP
Supplier:  Enzo Life Sciences BMLPI1280025
Encompass_Preferred
A novel substrate-based (γ-40 site) aldehyde inhibitor of γ-secretase. In whole cells overexpressing amloid ß protein precursor, Boc-GVV-CHO dose-dependently inhibited amyloid ß cleavage. γ-secretase is an important therapeutic target for strategies designed to lower amyloid ß production. Alternative name: Boc-GVV-CHO. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25 mg/ml) or ethanol (10 mg/ml warm). Long Term Storage: -20°C.
Catalog No. 50-201-1466